6-Nitro-1,2,3,4-tetrahydroquinoxaline ≥95%

Supplier: Matrix Scientific

074237-250MG 074237-5G
102535-372EA 400 USD
102535-372 102535-376
6-Nitro-1,2,3,4-tetrahydroquinoxaline ≥95%
6-Nitro-1,2,3,4-tetrahydroquinoxaline

Hazard: Irritant

Formula: C₈H₉N₃O₂
MDL Number: MFCD03788188
CAS Number: 41959-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR