3-(Chlorosulfonyl)benzoic acid ≥95%

Supplier: Matrix Scientific

074232-5G 074232-100G
102535-344EA 16.92 USD
102535-344 102535-348
3-(Chlorosulfonyl)benzoic acid ≥95%
3-(Chlorosulphonyl)benzoic acid

Hazard: Irritant

Formula: C₇H₅ClO₄S
MDL Number: MFCD00024877
CAS Number: 4025-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR