4-Fluoro-2-methoxybenzoic acid ≥95%

Supplier: Matrix Scientific

074229-1G 074229-10G
102535-326EA 193.85 USD
102535-326 102535-330
4-Fluoro-2-methoxybenzoic acid ≥95%
4-Fluoro-2-methoxybenzoic acid

Hazard: Irritant

Formula: C₈H₇FO₃
MW: 170.14
MDL Number: MFCD00673010
CAS Number: 395-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR