6-Chloro-5-nitro-o-cresol ≥95%

Supplier: Matrix Scientific

074227-5G 074227-100G
102535-316EA 284.62 USD
102535-316 102535-320
6-Chloro-5-nitro-o-cresol ≥95%
6-Chloro-5-nitro-o-cresol

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD03425888
CAS Number: 39183-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR