1-(4-Methoxyphenyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

074224-5G 074224-10G 074224-100G
102535-298EA 55.38 USD
102535-298 102535-300 102535-302
1-(4-Methoxyphenyl)piperazine dihydrochloride ≥95%
1-(4-Methoxyphenyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈Cl₂N₂O
MW: 265.18 g/mol
MDL Number: MFCD00012767
CAS Number: 38869-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR