1-(4-Chlorophenyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

074223-10G 074223-100G
102535-294EA 23.08 USD
102535-294 102535-296
1-(4-Chlorophenyl)piperazine dihydrochloride ≥95%
1-(4-Chlorophenyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₃N₂
MW: 269.60 g/mol
MDL Number: MFCD00012766
CAS Number: 38869-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR