5-Bromo-2-ethylpyridine ≥95%

Supplier: Matrix Scientific

074222-250MG 074222-1G 074222-5G
102535-288EA 498.46 USD
102535-288 102535-290 102535-292
5-Bromo-2-ethylpyridine ≥95%
5-Bromo-2-ethylpyridine

Hazard: Irritant

Formula: C₇H₈BrN
MDL Number: MFCD11226235
CAS Number: 38749-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR