3-Fluoro-2-iodobenzoic acid ≥95%

Supplier: Matrix Scientific

074221-1G 074221-5G
102535-284EA 138.46 USD
102535-284 102535-286
3-Fluoro-2-iodobenzoic acid ≥95%
3-Fluoro-2-iodobenzoic acid

Hazard: Irritant

Formula: C₇H₄FIO₂
MDL Number: MFCD07774249
CAS Number: 387-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR