6-Fluorosalicylaldehyde ≥95%

Supplier: Matrix Scientific

074219-1G 074219-5G 074219-10G
102535-272EA 284.62 USD
102535-272 102535-274 102535-276
6-Fluorosalicylaldehyde ≥95%
6-Fluorosalicylaldehyde

Hazard: Irritant

Formula: C₇H₅FO₂
MDL Number: MFCD01090996
CAS Number: 38226-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR