2-Amino-5-bromophenol ≥95%

Supplier: Matrix Scientific

074218-5G 074218-25G
102535-268EA 106.15 USD
102535-268 102833-674
2-Amino-5-bromophenol ≥95%
2-Amino-5-bromophenol

Hazard: Irritant/Light Sensitive

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Melting Pt: 135…140 °C (dec.)
MDL Number: MFCD03095028
CAS Number: 38191-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR