5-Bromo-3-chloro-2-fluoropyridine ≥95%
Supplier: Matrix Scientific
074217-1G
074217-5G
074217-25G
102535-260EA
36.92
USD
102535-260
102535-262
102551-878
5-Bromo-3-chloro-2-fluoropyridine ≥95%
5-Bromo-3-chloro-2-fluoropyridine
Hazard: Irritant
Formula:
C₅H₂BrClFN MW: 210.43 g/mol |
MDL Number:
MFCD09834365 CAS Number: 38185-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...