4-Chloro-3-nitrophenylacetic acid ≥95%

Supplier: Matrix Scientific

074214-1G 074214-5G 074214-10G
102535-246EA 355.38 USD
102535-246 102535-248 102535-250
4-Chloro-3-nitrophenylacetic acid ≥95%
4-Chloro-3-nitrophenylacetic acid

Hazard: Irritant

Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
MDL Number: MFCD00016990
CAS Number: 37777-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR