(R)-(Piperidin-3-yl)methanol ≥95%

Supplier: Matrix Scientific

074213-1G 074213-5G 074213-10G
102535-240EA 275.38 USD
102535-240 102535-242 102535-244
(R)-(Piperidin-3-yl)methanol ≥95%
(R)-(Piperidin-3-yl)methanol

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.1700 g/mol
MDL Number: MFCD09751036
CAS Number: 37675-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR