Methyl-4,6-dichloropyridazine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4,6-dichloropyridazine-3-carboxylate

074212-250MG 074212-1G
102535-236EA 372.69 USD
102535-236 102535-238
Methyl-4,6-dichloropyridazine-3-carboxylate ≥95%
Methyl-4,6-dichloropyridazine-3-carboxylate

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O₂
MW: 207.0142 g/mol
MDL Number: MFCD09953612
CAS Number: 372118-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR