4,6-Difluoro-m-toluic acid ≥95%
Supplier: Matrix Scientific
074211-1G
074211-5G
074211-10G
102535-230EA
533.85
USD
102535-230
102535-232
102535-234
4,6-Difluoro-m-toluic acid ≥95%
4,6-Difluoro-m-toluic acid
Hazard: Irritant
Formula:
C₈H₆F₂O₂ |
MDL Number:
MFCD09835285 CAS Number: 367954-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...