2-Bromo-4-(trifluoromethyl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(trifluoromethyl)benzonitile

074204-1G 074204-5G 074204-10G
102535-196EA 15.38 USD
102535-196 102535-198 102535-200
2-Bromo-4-(trifluoromethyl)benzonitrile ≥95%
2-Bromo-4-(trifluoromethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₃BrF₃N
MW: 250.0200 g/mol
MDL Number: MFCD06659467
CAS Number: 35764-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR