2,4-Dimethoxypyrimidine ≥95%

Supplier: Matrix Scientific

074203-5G 074203-10G 074203-100G
102535-190EA 178.46 USD
102535-190 102535-192 102535-194
2,4-Dimethoxypyrimidine ≥95%
2,4-Dimethoxypyrimidine

Hazard: Irritant

Formula: C₆H₈N₂O₂
MDL Number: MFCD00056028
CAS Number: 3551-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR