(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate, tert-Butyl L-Pyroglutamate, tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, tert-Butyl (S)-2-pyrrolidone-5-carboxylate

074202-1G 074202-5G 074202-10G
102535-184EA 143.08 USD
102535-184 102535-186 102535-188
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥95%
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD06659481
CAS Number: 35418-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR