4-Amino-2-chloro-5-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-nitro-4-(trifluoromethyl)aniline

074201-1G 074201-5G 074201-10G
102535-178EA 284.62 USD
102535-178 102535-180 102535-182
4-Amino-2-chloro-5-nitrobenzotrifluoride ≥95%
4-Amino-2-chloro-5-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₇H₄ClF₃N₂O₂
MW: 240.5700 g/mol
MDL Number: MFCD01312277
CAS Number: 35375-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR