5-Methoxyindol-3-ylacetic acid ≥95%

Supplier: Matrix Scientific

074199-1G 074199-5G 074199-10G
102535-166EA 249.23 USD
102535-166 102535-168 102535-170
5-Methoxyindol-3-ylacetic acid ≥95%
5-Methoxyindol-3-ylacetic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00005638
CAS Number: 3471-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR