4-Chloro-2-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

074198-5G 074198-10G 074198-100G
102535-160EA 72.31 USD
102535-160 102535-162 102535-164
4-Chloro-2-nitrobenzonitrile ≥95%
4-Chloro-2-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
MDL Number: MFCD00027398
CAS Number: 34662-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR