4-Fluoroisatin ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Fluoroindoline-2,3-dione

074197-1G 074197-5G
102535-156EA 326.15 USD
102535-156 102535-158
4-Fluoroisatin ≥95%
4-Fluoroisatin

Hazard: Irritant

Formula: C₈H₄FNO₂
MW: 165.1200 g/mol
Density: 1.477
MDL Number: MFCD01175824
CAS Number: 346-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR