4-Bromo-2-methylisoquinolin-1(2H)-one ≥95%

Supplier: Matrix Scientific

074194-250MG 074194-1G 074194-5G
102535-142EA 481.82 USD
102535-142 102535-144 102535-146
4-Bromo-2-methylisoquinolin-1(2H)-one ≥95%
4-Bromo-2-methylisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₁₀H₈BrNO
MDL Number: MFCD00489133
CAS Number: 33930-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR