4-Phenyl-1-butanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Phenylbutan-1-ol

074193-5G 074193-10G 074193-100G
102535-136EA 126.15 USD
102535-136 102535-138 102535-140
4-Phenyl-1-butanol ≥95%
4-Phenyl-1-butanol

Hazard: Irritant

Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 140
MDL Number: MFCD00002971
CAS Number: 3360-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR