3-Chloro-2-methoxybenzoic acid ≥95%
Supplier: Matrix Scientific
074189-1G
074189-5G
074189-10G
102535-112EA
107.69
USD
102535-112
102535-114
102535-116
3-Chloro-2-methoxybenzoic acid ≥95%
3-Chloro-2-methoxybenzoic acid
Hazard: Irritant
Formula:
C₈H₇ClO₃ |
MDL Number:
MFCD00179306 CAS Number: 3260-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...