2-(N-Methylamino)-3-hydroxymethylpyridine ≥95%

Supplier: Matrix Scientific

074187-1G 074187-5G
102535-104EA 83.08 USD
102535-104 102535-106
2-(N-Methylamino)-3-hydroxymethylpyridine ≥95%
2-(N-Methylamino)-3-hydroxymethylpyridine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
MDL Number: MFCD06254696
CAS Number: 32399-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR