4-Nitro-α,α,α-trifluoro-o-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-2-(trifluoromethyl)benzoic acid

074183-5G 074183-25G 074183-100G
102535-084EA 101.54 USD
102535-084 102551-874 102551-876
4-Nitro-α,α,α-trifluoro-o-toluic acid ≥95%
4-Nitro-α,α,α-trifluoro-o-toluic acid

Hazard: Irritant

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
Melting Pt: 135…137.5 °C
MDL Number: MFCD04039246
CAS Number: 320-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR