2-(5-Bromopyridin-2-yl)acetonitrile ≥95%

Supplier: Matrix Scientific

074178-1G 074178-5G
102535-054EA 90.77 USD
102535-054 102535-056
2-(5-Bromopyridin-2-yl)acetonitrile ≥95%
2-(5-Bromopyridin-2-yl)acetonitrile

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD07644625
CAS Number: 312325-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR