3-Methyl-4-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

074177-10G 074177-100G
102535-050EA 36.92 USD
102535-050 102535-052
3-Methyl-4-nitrobenzoic acid ≥95%

Purity: 95+% CAS: 3113-71-1 MDL Number: MFCD00007168 Molecular Formula: C8H7NO4 Molecular Weight: 181,15 Hazard: Irritant

Formula: C₈H₇NO₄
MDL Number: MFCD00007168
CAS Number: 3113-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR