3-Bromo-2-methylthiophene ≥95%

Supplier: Matrix Scientific

074174-1G 074174-5G 074174-10G
102535-034EA 126.15 USD
102535-034 102535-036 102535-038
3-Bromo-2-methylthiophene ≥95%
3-Bromo-2-methylthiophene

Hazard: Irritant

Formula: C₅H₅BrS
MW: 177.0622 g/mol
MDL Number: MFCD09033008
CAS Number: 30319-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR