Ethyl-1-methyl-1H-imidazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylimidazole-2-carboxylic acid ethyl ester, Ethyl 1-methylimidazole-2-carboxylate, Ethyl 1-methyl-1H-imidazole-2-carboxylate, Ethyl 1-Methylimidazole-2-carboxylate

074173-1G 074173-5G 074173-10G
102535-028EA 90.77 USD
102535-028 102535-030 102535-032
Ethyl-1-methyl-1H-imidazole-2-carboxylate ≥95%
Ethyl-1-methyl-1H-imidazole-2-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD02683326
CAS Number: 30148-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR