3,5-Dimethylisoxazole ≥95%

Supplier: Matrix Scientific

074172-10G 074172-100G
102535-024EA 44.62 USD
102535-024 102535-026
3,5-Dimethylisoxazole ≥95%
3,5-Dimethylisoxazole

Hazard: Irritant

Formula: C₅H₇NO
MW: 97.12 g/mol
MDL Number: MFCD00003156
CAS Number: 300-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR