(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥95%

Supplier: Matrix Scientific

Synonyms: (1S,2S)-(-)-1,2-Diphenylethylenediamine, (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, (S,S)-DPEN

074171-1G 074171-5G 074171-10G
102535-018EA 143.08 USD
102535-018 102535-020 102535-022
(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥95%
(1S,2S)-(-)-1,2-Diphenylethylenediamine

Hazard: Irritant

Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
MDL Number: MFCD00082751
CAS Number: 29841-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR