(R)-2-Hydroxy-4-phenylbutyric acid ≥95%

Supplier: Matrix Scientific

074170-1G 074170-5G 074170-10G
102535-012EA 87.69 USD
102535-012 102535-014 102535-016
(R)-2-Hydroxy-4-phenylbutyric acid ≥95%
(R)-2-Hydroxy-4-phenylbutyric acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00192219
CAS Number: 29678-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR