2,5-Dibromonicotinic acid ≥95%
Supplier: Matrix Scientific
074169-1G
074169-5G
102535-008EA
29.23
USD
102535-008
102535-010
2,5-Dibromonicotinic acid ≥95%
2,5-Dibromonicotinic acid
Hazard: Irritant
Formula:
C₆H₃Br₂NO₂ MW: 280.9000 g/mol |
MDL Number:
MFCD06254512 CAS Number: 29312-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...