4-Acetoxy-2-azetidinone ≥95%

Supplier: Matrix Scientific

074168-1G 074168-5G
102535-004EA 161.54 USD
102535-004 102535-006
4-Acetoxy-2-azetidinone ≥95%
4-Acetoxy-2-azetidinone

Hazard: Irritant

Formula: C₅H₇NO₃
MDL Number: MFCD00010593
CAS Number: 28562-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR