2-Amino-5-chlorophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-hydroxyaniline

074167-5G 074167-10G 074167-100G
102534-998EA 90.77 USD
102534-998 102535-000 102535-002
2-Amino-5-chlorophenol ≥95%
2-Amino-5-chlorophenol

Hazard: Irritant

Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD02093863
CAS Number: 28443-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR