2-Nitro-4-aminodiphenylamine ≥95%
Supplier: Matrix Scientific
074164-1G
074164-5G
074164-10G
102534-980EA
107.69
USD
102534-980
102534-982
102534-984
2-Nitro-4-aminodiphenylamine ≥95%
2-Nitro-4-aminodiphenylamine
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ MW: 229.24 g/mol |
MDL Number:
MFCD00239471 CAS Number: 2784-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...