2-Amino-4-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-5-methylaniline

074162-1G 074162-5G 074162-10G
102534-968EA 32.31 USD
102534-968 102534-970 102534-972
2-Amino-4-methylbenzonitrile ≥95%
2-Amino-4-methylbenzonitrile

Hazard: Toxic/Stored under Argon/Air Sensitive

Formula: C₈H₈N₂
Melting Pt: 91-93°C
MDL Number: MFCD00173706
CAS Number: 26830-96-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR