(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Bromo-2-hydroxy-2-methylpropanoic acid, (R)-3-Bromo-2-hydroxy-2-methylpropionic acid

074161-1G 074161-5G
102534-964EA 100 USD
102534-964 102534-966
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid ≥95%
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid

Hazard: Irritant

Formula: C₄H₇BrO₃
MW: 183.0006 g/mol
Melting Pt: 109…113 °C
MDL Number: MFCD09751022
CAS Number: 261904-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR