1-Boc-3,3-dimethylpiperazine ≥95%

Supplier: Matrix Scientific

074158-250MG 074158-1G 074158-5G
102534-946EA 250.11 USD
102534-946 102534-948 102534-950
1-Boc-3,3-dimethylpiperazine ≥95%
1-Boc-3,3-dimethylpiperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD07371651
CAS Number: 259808-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR