3-Nitro-4-(trifluoromethyl)phenol ≥95%
Supplier: Matrix Scientific
074157-1G
074157-5G
074157-10G
102534-940EA
569.23
USD
102534-940
102534-942
102534-944
3-Nitro-4-(trifluoromethyl)phenol ≥95%
3-Nitro-4-(trifluoromethyl)phenol
Hazard: Irritant
Formula:
C7H4F3NO3 MW: 207.1100 g/mol |
MDL Number:
MFCD07783660 CAS Number: 25889-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...