(R)-2-Amino-1-phenylethanol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(-)-2-Amino-1-phenylethanol, (R)-(-)-alpha-(Aminomethyl)benzyl alcohol

074155-1G 074155-5G
102534-930EA 146.15 USD
102534-930 102534-932
(R)-2-Amino-1-phenylethanol ≥95%
(R)-2-Amino-1-phenylethanol

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₁NO
Melting Pt: 57…63 °C
MDL Number: MFCD00239406
CAS Number: 2549-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR