2-Hydroxy-4-(trifluoromethyl)quinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Trifluoromethyl-2(1H)-quinolinone

074152-1G 074152-5G
102534-916EA 640 USD
102534-916 102534-918
2-Hydroxy-4-(trifluoromethyl)quinoline ≥95%
2-Hydroxy-4-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
MDL Number: MFCD03407380
CAS Number: 25199-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR