4-Chloroindole-3-acetic acid ≥95%

Supplier: Matrix Scientific

074151-1G 074151-5G 074151-25G
102534-910EA 246.15 USD
102534-910 102534-912 102663-982
4-Chloroindole-3-acetic acid ≥95%
4-Chloroindole-3-acetic acid

Hazard: Irritant

Formula: C₁₀H₈ClNO₂
Melting Pt: 184…187 °C
MDL Number: MFCD00216155
CAS Number: 2519-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR