4'-Amino-2'-hydroxyacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Amino-2-hydroxyphenyl)ethanone, 1-(4-Amino-2-hydroxyphenyl)ethan-1-one

074150-1G 074150-5G
102534-906EA 156.92 USD
102534-906 102534-908
4'-Amino-2'-hydroxyacetophenone ≥95%
4'-Amino-2'-hydroxyacetophenone

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
MDL Number: MFCD00100636
CAS Number: 2476-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR