4,6-Difluoroindole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074149-1G 074149-5G
102534-902EA 533.85 USD
102534-902 102534-904
4,6-Difluoroindole-2-carboxylic acid ≥95%
4,6-Difluoroindole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅F₂NO₂
MDL Number: MFCD01863158
CAS Number: 247564-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR