6-Hydroxy-1-naphthoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-1-naphthalenecarboxylic acid

074146-5G 074146-100G
102534-886EA 230.77 USD
102534-886 102534-890
6-Hydroxy-1-naphthoic acid ≥95%
6-Hydroxy-1-naphthoic acid

Hazard: Irritant

Formula: C₁₁H₈O₃
MW: 188.1800 g/mol
MDL Number: MFCD00209983
CAS Number: 2437-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR