2-Chloro-4-(trifluoromethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-trifluoromethylbenzoic acid, 2-Chloro-α,α,α-trifluoro-p-toluic acid

074142-1G 074142-5G
102534-864EA 27.69 USD
102534-864 102534-866
2-Chloro-4-(trifluoromethyl)benzoic acid ≥95%
2-Chloro-4-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Melting Pt: 114-117°C
MDL Number: MFCD06208178
CAS Number: 23228-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR