3-Bromo-4-hydroxybenzonitrile ≥95%
Supplier: Matrix Scientific
074141-5G
074141-10G
074141-100G
102534-858EA
107.69
USD
102534-858
102534-860
102534-862
3-Bromo-4-hydroxybenzonitrile ≥95%
3-Bromo-4-hydroxybenzonitrile
Hazard: Irritant
Formula:
C₇H₄BrNO MW: 198.02 g/mol |
MDL Number:
MFCD00143096 CAS Number: 2315-86-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...